missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBN Mouse anti-Human, Unconjugated, Clone: 1E11E10, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | NBN |
---|---|
Clone | 1E11E10 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829325
|
Proteintech
66980-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819325
|
Proteintech
66980-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NBN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NBN. This antibody recognizes human antigen. The NBN antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
NBN | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
4683 | |
Primary |
1E11E10 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG2a |