Learn More
Invitrogen™ Neon™ Transfection System Pipette Station
Click to view available options
:
1 each
Description
The Neon™ Transfection System Pipette Station is designed specifically for use with the Neon™ Transfection System device (MPK5000). Each device (MPK5000) already comes with a pipette station. The pipette station is also sold separately for those labs needing and additional pipette station or replacement for their existing pipette station.
For more information on the product, please click here
Specifications
Specifications
| Content And Storage | The Neon™ Transfection System Pipette station comes with 1 x pipette station. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions. |
| For Use With | Transfection Grade Nucleic Acid Purification Kits, siRNA Vectors, DNA Vectors |
| Product Line | Neon |
| Type | Electroporation Pipette Tip |
| For Use With (Equipment) | Neon™ Transfection System |
| Quantity | 1 each |
| Serum Compatible | Yes |
| Shipping Condition | Room Temperature |
| Starting Material Cell No. | 2x10^4 - 6 x10^8 cells; Varies by cell type |
| System Type | Neon™ Transfection System |