missing translation for 'onlineSavingsMsg'
Learn More

Nepicastat hydrochloride, MedChemExpress

Product Code. 16468317
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16468317

Brand: Medchem Express HY13289A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nepicastat hydrochloride (SYN-117 hydrochloride) is a selective, potent, and orally active inhibitor of dopamine-beta-hydroxylase. Nepicastat hydrochloride produces concentration-dependent inhibition of bovine (IC50=8.5 nM) and human (IC50=9 nM) dopamine-beta-hydroxylase. Nepicastat hydrochloride can cross the blood-brain barrier (BBB).

Nepicastat hydrochloride (SYN-117 hydrochloride) is a selective, potent, and orally active inhibitor of dopamine-beta-hydroxylase. Nepicastat hydrochloride produces concentration-dependent inhibition of bovine (IC50=8.5 nM) and human (IC50=9 nM) dopamine-beta-hydroxylase. Nepicastat hydrochloride can cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material Nepicastat hydrochloride
CAS 170151-24-3
Color White
Purity Grade Notes Research
Molecular Formula C14H16ClF2N3S
Quantity 5 mg
Synonym SYN-117 hydrochloride RS-25560-197 hydrochloride
Solubility Information DMSO : 6 mg/mL (18.08 mM; Need ultrasonic) ∣H2O : 2 mg/mL (6.03 mM; Need ultrasonic and warming)
SMILES FC1=CC(F)=C2CC[C@@H](CC2=C1)N3C(CN)=CNC3=S.Cl
Molecular Weight (g/mol) 331.81
Formula Weight 331.81
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.