missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-2 Rabbit anti-Human, Mouse, Rat, Clone: 8F2Q8, Novus Biologicals™
Rabbit Monoclonal Antibody
196.00 EUR - 494.00 EUR
Specifications
Antigen | Neurogenin-2 |
---|---|
Clone | 8F2Q8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18340893
|
Bio-Techne
NBP3-15924-20UL |
20 μg |
196.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331287
|
Bio-Techne
NBP3-15924-100UL |
100 μg |
494.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin-2 Monoclonal antibody specifically detects Neurogenin-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Neurogenin-2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
63973 | |
IgG | |
Affinity purified |
8F2Q8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Neurogenin-2 (Q9H2A3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title