Learn More
Eppendorf™ Scientific Test Tube Racks
Adjustable angle and tube height
366.00 EUR
Product Code | Brand | Size | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Diameter (Metric) | Price | Quantity & Availability | ||||
11399924
|
Eppendorf™
M1289-0002 |
Small | 12 to 15 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11389924
|
Eppendorf™
M1289-0001 |
Small | 8 to 11 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11309934
|
Eppendorf™
M1289-0003 |
Small | 15 to 18 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11319934
|
Eppendorf™
M1289-0004 |
Small | 18 to 21 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11329934
|
Eppendorf™
M1289-0005 |
Small | 22 to 26 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11329924
|
Eppendorf™
M1289-0010 |
Medium | 8 to 11 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11339924
|
Eppendorf™
M1289-0020 |
Medium | 12 to 15 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11349924
|
Eppendorf™
M1289-0030 |
Medium | 15 to 18 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11339934
|
Eppendorf™
M1289-0006 |
Small | 26 to 30 mm |
366.00 EUR
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
||||
11369924
|
Eppendorf™
M1289-0050 |
Medium | 22 to 26 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11359924
|
Eppendorf™
M1289-0040 |
Medium | 18 to 21 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
11379924
|
Eppendorf™
M1289-0060 |
Medium | 26 to 30 mm |
366.00 EUR
Each |
In Stock
Log in to see stock. |
||||
Description
- Inserts to help retain tubes and prevent them from spinning in the rack
- Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
- Stainless-steel
Specifications
Test Tube Rack | |
Stainless Steel |