Learn More
Thermo Scientific™ CellSensor™ NFAT-bla RA-1 Cell Line
B cell receptor (BCR)-mediated signaling pathways are important for B cell proliferation and differentiation. Abnormal B cell signaling has been linked to various diseases such as lupus, lymphoma, and various immune disorders.
Brand:  Thermo Scientific™ K1490
Product Code. 10318824
  • 11130.00 EUR / Each
Estimated Shipment: 23-10-2024
Description
B cell receptor (BCR)-mediated signaling pathways are important for B cell proliferation and differentiation. Abnormal B cell signaling has been linked to various diseases such as lupus, lymphoma, and various immune disorders. Antigen binding to the BCR promotes the activation of several protein tyrosine kinases (PTK), which leads to phosphorylation of the BCR complex and recruitment and activation of the PTK Syk. This in turn promotes phosphorylation of PLCγ, Shc, and Vav. Additionally, the Tec family member Btk is recruited to the plasma membrane, where it is involved in activation of PLCγ. Initiation of B lymphocyte activation is dependent on the tyrosine phosphorylation-dependent formation of multimolecular effector protein complexes that activate downstream signaling pathways including PKC/Ca2+/NFAT. This cell-based assay has been thoroughly tested and validated by Invitrogen and is suitable for immediate use in screening applications. The following information illustrates the comprehensive assay testing completed and validation of assay performance under optimized conditions (Figure 1).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell-based beta lactamase reporter gene
CellSensor™ NFAT-bla RA-1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Signaling Pathway
1 Vial
Cell Line
CellSensor™
Dividing Cells
Fluorescence
CellSensor™
SYK, PKC and calcium pathway, BTK
RA-1
Signaling Pathway