Learn More
Nickel boride, 99% (metals basis), Thermo Scientific Chemicals
Description
Nickel boride is an efficient catalyst and reducing agent. It is used as a heterogeneous hydrogenation catalyst. It can also be used to cleave thioacetals. It is used in a protection-deprotection scheme for triterpenoid ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White to Gray |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN3077 |
Formula Weight | 69.52 |
Percent Purity | 99% |
Odor | Odorless |
Density | 7.39g/mL |
Assay | (metals basis) |
Chemical Name or Material | Nickel boride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.