Learn More
WTW™ Nickel Reagent Test Kit
With certificate for optimum precision.
Brand: WTW™ 250443
This item is not returnable.
View return policy
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Barbituric acid-pyridine Carboxylic Acid | |
For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers | |
250 | |
Photometric |
For waste water, drinking water, surface water | |
Certificates | |
0.02 - 5.00mg/I Ni | |
Photometric test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- nickel test method
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : iodine