Learn More
WTW™ Nitrate Test Kits
Brand: WTW™ 250411
This item is not returnable.
View return policy
Description
Specifications
Phenol derivative | |
PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
45 | |
Photometric |
For waste water, drinking water, surface water | |
Certificates | |
25 Tests | |
CombiCheck Nitrate (NO3) cell test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- nitrate cell test in seawater
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, phosphoric acid