missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 65% w/w, Extra Pure, Scharlab

Product Code. 11350456
Click to view available options
Quantity:
5 L
Packaging:
HDPE Carboy
Unit Size:
5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11350456

Brand: Scharlab AC1599005P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Suitable for qualitative and semi-quantitative work. Nitric Acid, 65% w/w, Extra Pure, Scharlab is often used in organic chemistry and the pharmaceutical industry for purifying synthesized products when higher grade solvents than Synthesis Grade are required.

Extra Pure grade specifications are less strict than Reagent Grade, but are significantly higher than industrial products. They are often used in the pharmaceutical industry, either as a reagent or as raw materials for production.

Specifications

CAS 7697-37-2
Melting Point -32°C
Boiling Point 122°C
Density 1.41g/cm3
Heavy Metals (as Pb) Max. 0.00005%
Packaging HDPE Carboy
Residue after Ignition Max. 0.0005%
Vapor Pressure 9.4 hPa
Assay Percent Range 64-66%
Quantity 5 L
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
UN Number 2031
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Solubility Information Miscible in water
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01g/mol
Grade Applications: oxidizing agent, synthesis of nitrates and organic nitro compounds, in pharma industry
Assay w/w
Chemical Name or Material Nitric acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.