Learn More
WTW™ Nitrite Test Kits
Brand:  WTW™ 250445
Product Code. 10772752
  • 261.00 EUR / Each
Estimated Shipment: 22-01-2025
Specifications
Chromosulfuric Acid oxidation
For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
1000
For waste water, ground and surface water, drinking water, sea water, food and soils after corresponding sample preparation
Certificates
Photometric
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • nitrate test method: photometric
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : sulphanilic acid. N-(1-napjtl)ethylenediamine dihydrochloride