Learn More
WTW™ Nitrite Test Kits
Brand: WTW™ 250445
This item is not returnable.
View return policy
Description
Specifications
Chromosulfuric Acid oxidation | |
For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
1000 |
For waste water, ground and surface water, drinking water, sea water, food and soils after corresponding sample preparation | |
Certificates | |
Photometric |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- nitrate test method: photometric
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sulphanilic acid. N-(1-napjtl)ethylenediamine dihydrochloride