Learn More
Norell™ Ultra Precision Borosilicate Glass NMR Sample Tubes
Product Code. p-8013337
Click to view available options
Length (Metric):
178 mm
203 mm
Diameter (Metric) Inner:
2.41 mm
4.2 mm
Camber (Metric):
0.006 mm
0.013 mm
0.019 mm
0.025 mm
0.035 mm
0.038 mm
Concentricity (Metric):
0.0040 mm
0.0050 mm
0.0070 mm
0.0100 mm
Unit Size:
Pack of 5
Product Code. 12880036
Brand: Norell™ S34007
Only null left
Add to basket
Add to basket
For auto-sampling systems and high-throughput NMR
  • Specifically designed for high-resolution NMR, tubes in the Select Series™ are manufactured from ASTM Type 1 Class A borosilicate glass
  • Whether spinning or non-spinning, doing structure determination of small molecules or determining the 3D structure of a membrane protein, the Norell™ Select Series™ of NMR tubes offers unsurpassed performance, reproducibility and reliability compared to other brands
  • You should see an immediate improvement in resolution when calculating i-coupling constants or acquiring data close to your spectrophotometer noise level, resulting in improved peak symmetry and shimmering times
  • In turn this leads to shorter analysis times and improved sample throughput
  • There are also no worries with freeze/thaw degassing or high temperature studies, these tubes can tolerate thermal shock up to 200°C
  • All tubes have polished ends making them less susceptible to breaking
  • Colored end caps are included
Specifications
Camber (Metric) 0.019 mm
Concentricity (Metric) 0.0070 mm
Length (Metric) 178 mm
Material Borosilicate Glass
Product Type NMR Sample Tube
Packaging 5 Pack
Diameter (Metric) Outer 2.99 mm
Diameter (Metric) Inner 2.41 mm
Includes Caps