Learn More
GPE Scientific™ Norell™ Thin-Wall Transparent Fluoropolymer FEP Tubing
Manufactured from special virgin grade thermoplastic FEP fluoropolymer
Brand:  GPE Scientific™ TWT-3
 View more versions of this product
Product Code. 13110833
  • 119.00 EUR / Pack of 4
Estimated Shipment: 22-04-2025
Description
  • Unlike PTFE tubing, this material can be heat sealed
  • Heat sealing can offer extremely large possibilities for packaging and/or storage of samples (corrosives, liquids, solids, etc.)
  • Fluoropolymer tubing can also be used where snug and tight fit over glass or metal tubing is required
Specifications
Specifications
3.5 mm