missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Novex™ Tris-Glycine Plus Midi Protein Gels, 10%, 1.0 mm

Novex Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based on traditional Laemmli protein electrophoresis technology, allowing the use of Laemmli sample and running buffers.

223.00 EUR - 235.00 EUR

Specifications

Gel Percentage 10%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Tris-Glycine Plus
Separation Range 30 to 250 kDa
View More Specs

Products 5
Product Code Brand Wells Price Quantity & Availability  
Product Code Brand Wells Price Quantity & Availability  
15932354
View Documents
Novex™
WXP01012BOX
12 + 2-well
223.00 EUR
Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
15743488
View Documents Promotion Details
Novex™
WXP01020BOX
20-well
223.00 EUR
Pack of 10
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
15959326
View Documents
Novex™
WXP01020BOXA
20-well, w/adapters
235.00 EUR
Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
15942354
View Documents
Novex™
WXP01026BOX
26-well
223.00 EUR
Pack of 10
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
16346585
View Documents
Novex™
WXP01026BOXA
26-well, w/adapters
235.00 EUR
Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
Description

Description

Novex Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based on traditional Laemmli protein electrophoresis technology, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus Midi gels are a 1.0-mm thick, wider format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples.

Features of Novex Tris-Glycine Plus gels include:
Improved shelf life—store gels for up to 12 months at 4°C
Flexibility—use for native and denaturing protein assays
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine Plus Midi gels come in fixed concentrations of 10% and 12%, as well as gradient concentrations of 4–12%, 4–20%, and 8–16%. Select from our many well formats, including 12+2-, 20-, and 26-well.

Run your proteins in native or denatured form
Novex Tris-Glycine Plus gels do not contain SDS and so can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26754). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672). The gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or conveniently with the Bio-Rad Criterion Cell using our adapters (WA0999).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) when performing traditional wet transfer. Rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Specifications

Specifications

10%
1.0 mm
30 to 250 kDa
Molecular Weight
10 gels (1 box)
Store at 2°C to 8°C. Do not freeze.
13 cm
Midi
Tris-Glycine Plus
Denaturing, Native
Novex™
Wet Ice
8 cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.