missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tropifluor™ PVDF Membrane, 0.45 μm pore size, 15 cm X 15 cm

Provide optimized results in western blotting applications with dioxetane-based substrate solutions (CSPD™ or CDP-Star™ substrates)

Brand:  Invitrogen™ T2234

Product Code. 12614345

  • 227.00 EUR / Each
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provides the high binding and retention capacity that characterizes PVDF (polyvinylidene fluoride) membranes and high mechanical strength that permits convenient handling and withstands multiple re-probing. Low background and resulting superior signal-to-noise performance obtained with this membrane enables picogram detection levels of proteins in immunoblotting applications. Yields low background levels with use of Tropix chemiluminescent substrates and secondary antibody conjugates, enabling ultrasensitive detection. Recommended for immunoblotting applications where sensitivity is important.

  • Pore Size of 0.45 microns
  • Binding capacity: 125μg of protein/cm2
  • Size: 15 x 15cm

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

Services

Guaranteed for 12 months from the date of purchase.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 cm
5 sheet(s)
PVDF
Membranes may be stored at room temperature.
15 cm x 15 cm
15 cm
Sheet
0.45 μm
Room Temperature
Tropifluor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.