Learn More
Thermo Scientific™ NP-40 lysis buffer with glycerol (2X)
Product Code. 15422969
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
Product Code. 15422969
Brand: Thermo Scientific™ J60143.=E
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Concentration 2X
Content And Storage Keep cold
Format Viscous Liquid
Quantity 125 mL