missing translation for 'onlineSavingsMsg'
Learn More

NPE-caged-proton, Tocris Bioscience™

Product Code. 18095792
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Caged proton

NPE-caged-proton is a 1-(2-nitrophenyl)ethyl caged proton that releases a proton and a sulfate ion upon photolysis at 350 - 355 nm. Generates rapid acidifications (pH jumps) down to pH 2.

Specifications

CAS 1186195-63-0
Molecular Formula C8H8NNaO6S
Quantity 10 mg
Synonym 1-(2-Nitrophenyl)ethyl sulfate sodium salt
InChI Key LCCXAXUWDTVOHF-FYZOBXCZSA-M
SMILES CC(C1=CC=CC=C1[N+](=O)[O-])OS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;[(1R)-1-(2-nitrophenyl)ethyl] sulfate
Molecular Weight (g/mol) 269.203
PubChem CID 124081034
Formula Weight 269.21
Percent Purity >98%
Chemical Name or Material NPE-caged-proton
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.