missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRBF2 Monoclonal Antibody (K40009_14D6), Invitrogen™
Mouse Monoclonal Antibody
480.00 EUR - 846.00 EUR
Specifications
| Antigen | NRBF2 |
|---|---|
| Clone | K40009_14D6 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30198099
|
Invitrogen™
MA556173 |
200 μg |
846.00 EUR
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30194412
|
Invitrogen™
MA555793 |
50 μg |
480.00 EUR
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GQSPLSQKYS PSTEKCLPEI QGIFDRDPDT LLYLLQQKSE PAEPCIGSKA PKDDKTIIEE QATKIADLKR HVEFLVAENE RLRKENKQLK AEKARLLKGP IEKELDVDAD
NRBF2 Monoclonal antibody specifically detects NRBF2 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| NRBF2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR; COPR1; COPR2; Nrbf2; NRBF-2; nuclear receptor binding factor 2; nuclear receptor binding factor-2; nuclear receptor-binding factor 2 | |
| Recombinant human NRBF2 (immunogen range is 111-220). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Nrbf2 |
| K40009_14D6 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96F24 | |
| 29982 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title