missing translation for 'onlineSavingsMsg'
Learn More

NS-638, MedChemExpress

Product Code. 16454606
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454606

Brand: Medchem Express HY101428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NS-638 is a small nonpeptide molecule with Ca2+-channel blocking properties. K+-stimulated intracellular Ca2+-elevation is blocked with an IC50 value of 3.4 μM.

NS-638 is a small nonpeptide molecule with Ca2+-channel blocking properties. K+-stimulated intracellular Ca2+-elevation is blocked with an IC50 value of 3.4 μM.

Specifications

Chemical Name or Material NS-638
CAS 150493-34-8
Color White
Purity Grade Notes Research
Molecular Formula C15H11ClF3N3
Quantity 100 mg
Solubility Information DMSO : ≥ 34 mg/mL (104.38 mM)
SMILES NC1=NC2=CC(C(F)(F)F)=CC=C2N1CC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 325.72
Formula Weight 325.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.