Learn More
NS-638, MedChemExpress
Product Code. 16499886
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16499886
Brand: Medchem Express HY101428
MedChemExpress NS-638 is a small nonpeptide molecule with Ca2+-channel blocking properties. K+-stimulated intracellular Ca2+-elevation is blocked with an IC50 value of 3.4 μM.
NS-638 is a small nonpeptide molecule with Ca2+-channel blocking properties. K+-stimulated intracellular Ca2+-elevation is blocked with an IC50 value of 3.4 μM.
Specifications
Chemical Name or Material NS-638
CAS 150493-34-8
Color White
Purity Grade Notes Research
Molecular Formula C15H11ClF3N3
Quantity 50 mg
Solubility Information DMSO : ≥ 34 mg/mL (104.38 mM)
SMILES NC1=NC2=CC(C(F)(F)F)=CC=C2N1CC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 325.72
Formula Weight 325.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation