Learn More
Showa Best Gloves™ NSK26™ Coated Nitrile Gloves
SHOWA™ NSK26™ coated nitrile gloves protect the hand and arm from a wide range of chemical splashes whilst providing comfort and touch sensitivity
Brand:  Showa Best Gloves™ NSK26-11XL
 View more versions of this product
Product Code. 12910785
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Fully coated nitrile glove that absorbs perspiration for ultra-comfortable extended wear and with good mechanical resistance
  • Cut and sewn construction designed for easy movement and continuous wear
  • Protects the hand from oils, hydrocarbons, grease and abrasion, with effective long lasting textured grip
  • Extended protection on the forearm
  • Impermeable for working in damp or greasy environments
  • Surface provides tactile feel and better grip
  • Lower allergy risks
  • Suitable for use in agriculture, construction, food handling, forestry & fishing, manufacturing, municipal services, oil and gas, wet work
Specifications
Specifications
EN 374, EN388, FOOD CONTACT DIRECTIVE 2002 / 72 EC
Rough
Powder Free
Non-sterile
Elasticated
650 mm
Blue
Nitrile
11
Agrochemicals, chemical handling, paints, oils, industrial tool maintenance, refining, waste and water treatment
Cotton
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Sonderangebote
Sonderangebote