missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NU 7441

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NU 7441 is a potent and selective DNA-PK inhibitor (IC50 = 14 nM). Selective for DNA-PK over a range of kinases including mTOR, PI 3-K, ATM and ATR. Potentiates the effects of doxorubicin (Cat. No. 2252) and etoposide (Cat. No. 1226) in vitro and etoposide in vivo. Also enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency 2 to 3-fold, and decreases nonhomologous end-joining (NHEJ) frequency ∽40%.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | NU 7441 |
| Purity | >99% |
| Molecular Weight (g/mol) | 413.49 |
| Inhibitors | NU 7441 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction