Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Shop All Macherey-Nagel Products
Click to view available options
Unit Size:
Each
Material:
Nucleogel SAX 1000-8
Specifications
Specifications
| For Use With (Application) | HPLC, Separation of Sugars, Alcohols and Organic Acids |
| Solvent Compatibility | 0.01 N H2SO4 |
| Particle Size | 10 μm |
| Type | HPLC Column |