Learn More
Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates
Click to view available options
Pore Size:
0.4 μm
3 μm
8 μm
Dish Type:
12-Well Plate
24-Well Plate
6-Well Plate
Culture Area:
0.47 cm2
1.13 cm2
3.14 cm2
4.1 cm2
Unit Size:
Pack of 24
Pack of 48
Description
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Features:
- Thermo Scientific™ Nunclon™ Delta-treated and tested for cell attachment
- Tabs provide better grip and easy handling
- Nontoxic and nonpyrogenic
Ordering Information:
Packed in resealable bags
Specifications
Specifications
| Description | Polycarbonate Cell Culture Inserts with 3 micron pore size in 6-well plate |
| Inserts per Plate | 6 |
| Culture Area | 3.14 cm2 |
| Material | Polycarbonate (Membrane) |
| Volume (Metric) Working | 1.5 mL |
| Pore Density | <1.7 x 106 pores/cm2 |
| Pore Size | 3 μm |
| Type | Culture Insert |
| For Use With | Multidish 6 |
| No. per Pack | 6 |
| Show More |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.