missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUT Antibody (1G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-31146SS
This item is not returnable.
View return policy
Description
NUT Monoclonal specifically detects NUT in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NUT | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 2 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 2-5 ug/ml | |
C15orf55, chromosome 15 open reading frame 55, DKFZp434O192, FAM22H, MGC138683, MGC138684, Nuclear protein in testis, NUT, protein NUT | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1G6 | |
Unconjugated | |
Q86Y26 | |
NUTM1 | |
Partial recombinant protein made to an internal portion of human NUT (between amino acids 400-700) [UniProt Q86Y26]. | |
0.025 mg | |
Cancer | |
256646 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction