missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NVL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555801
This item is not returnable.
View return policy
Description
Immunogen sequence: VNRVLMKLQE QQKKNPEMED LPSKGVQEER LGTEPTSETQ DELQRLLGLL RDQDPLSEEQ MQGLCIELND FIVALSSVQP SAKREGFVTV PN Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 79%.
Gasdermin D is a member of the gasdermin family. Members of this family appear to play a role in regulation of epithelial proliferation. Gasdermin D has been suggested to act as a tumor suppressor. Alternatively spliced transcript variants have been described.Specifications
NVL | |
Polyclonal | |
Unconjugated | |
NVL | |
1200009I24Rik; Nuclear valosin-containing protein-like; nuclear VCP-like; nuclear VCP-like protein; Nvl; NVL2; NVLp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15381 | |
NVL | |
Recombinant protein corresponding to Human NVL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only