Learn More
Masterflex™ Nylon Threaded Adapter
Threaded fittings offer a more permanent installation. Available in a variety of materials and configurations.
Brand:  Masterflex™ 30401-34
Product Code. 11794949
  • 33.80 EUR / Pack of 10
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Description
  • Nylon threaded adapter
  • Use with rigid tubing for high-pressure applications
  • Adapts from a NPT male thread to a hose barb
  • Available in packs of ten
Specifications
Specifications
Threaded Adapter
1 in.
1 in. NPT(M) x 1 in. tubing ID
1 in. NPT(M)
Nylon
1 in.
Ethylene Oxide