Learn More
Brady™ Nylon Ties
Click to view available options
Color:
Blue
Orange
Purple
Red
White
Yellow
Unit Size:
1 pack
Description
- Cross-laminated polyethylene
- Matte finish
Specifications
Specifications
| Type | Ties |
| Material | Nylon |
| Color | Orange |
| Width (Metric) | 3.6 mm |
| Quantity | 100/Pk. |
| For Use With (Application) | Ideal for color-coding hard to mark items |