missing translation for 'onlineSavingsMsg'
Learn More
o-Cresolphthalein complexone tetrasodium salt, Thermo Scientific Chemicals Product Code.: 11419076

o-Cresolphthalein complexone tetrasodium salt, Thermo Scientific Chemicals

Product Code. 11419076
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11419076

Brand: Thermo Scientific Alfa Aesar A17941.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62698-54-8
Molecular Formula C32H28N2Na4O12
Molecular Weight (g/mol) 724.54
MDL Number MFCD00066371
InChI Key KSMPPVYJZJBIES-UHFFFAOYSA-J
Synonym phthalein complexon sodium salt, o-cresolphthalein complexone tetrasodium salt, sodium 2,2',2,2'-3-oxo-1,3-dihydroisobenzofuran-1,1-diyl bis 2-hydroxy-3-methyl-5,1-phenylene bis methylene bis azanetriyl tetraacetate, phthaleincomplexonsodiumsalt, phthalein complexone sodium salt, glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, sodium salt 1:4, glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, tetrasodium salt, tetrasodium 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-methylbenzene-3,1-diyl methanediylnitrilo tetraacetate, tetrasodium 2-5-1-3-bis carboxylatomethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl carboxylatomethyl amino acetate, tetrasodium 2-5-1-3-bis 2-oxido-2-oxoethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl-2-oxido-2-oxoethyl amino acetate
PubChem CID 112916
IUPAC Name tetrasodium;2-[[5-[1-[3-[[bis(carboxylatomethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1

Specifications

Quantity 1 g
Formula Weight 724.54
Chemical Name or Material o-Cresolphthalein complexone tetrasodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237511 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17941.03,A17941.06
Lot Number10237976 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17941.03,A17941.06
Lot NumberG4567A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17941.03,A17941.06
Lot Number10229968 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17941.03,A17941.06
Lot Number8239 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17941.03,A17941.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
o-Cresolphthalein complexone tetrasodium salt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.