missing translation for 'onlineSavingsMsg'
Learn More

o-Phthalaldehyde, MP Biomedicals™

Product Code. 11455985
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for precolumn derivatization of amino acids and other biochemical applications. o-Phthalaldehyde, MP Biomedicals is a dialdehyde, consisting of two formyl (CHO) groups attached to adjacent carbon centers on a benzene ring.

o-Phthalaldehyde is used for precolumn derivatization of amino acids for HPLC separation and for flow cytometric measurements of protein thiol groups. Used for fluorometric determination of histamine, histidine and other amino acids. Also used for cholesterol assay in the picomole range.

TRUSTED_SUSTAINABILITY

Specifications

CAS 643-79-8
Melting Point 53°C to 58°C
Molecular Formula C8H6O2
MDL Number MFCD00003335
Quantity 10 g
Synonym o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
Solubility Information Soluble in acetic acid (40mg/mL) or methanol (21mg/mL, clear solution); slightly soluble in water (5mg/mL).
InChI Key ZWLUXSQADUDCSB-UHFFFAOYSA-N
SMILES O=CC1=CC=CC=C1C=O
IUPAC Name benzene-1,2-dicarbaldehyde
Molecular Weight (g/mol) 134.13
PubChem CID 4807
ChEBI CHEBI:70851
Formula Weight 134.13
Percent Purity ≥98%
Physical Form Crystals
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'