Learn More
DWK Life Sciences O-ring, for stopcock keys with groove
For securing the DURAN™ valve plug with groove in the DURAN™ single way stopcock.
0.35 EUR - 32.40 EUR
Specifications
For Use With (Application) Sealing
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15991501
View Documents
DWK Life Sciences
286031408
24.0 NS
29.45 EUR
Pack of 50
In Stock
to see stock.
Add to basket
15814518
View Documents
DWK Life Sciences
286031605
29.2 NS
32.40 EUR
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15971501
View Documents
DWK Life Sciences
286030103
10.0 NS
10.30 EUR
Pack of 50
In Stock
to see stock.
Add to basket
10354961
View Documents
DWK Life Sciences
286031202
21.5 NS This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
The O-ring in the groove of the DURAN™ tap plug provided for this purpose secures the tap plug in the disposable tap.
  • Secures the valve plug from falling out of the valve
Specifications
Specifications
Sealing