Learn More
Motic™ Objective Lens for SMZ160
The Objective Lens is compatible with the SMZ-160 microscope.
Brand:  Motic™ 1101001708281
 View more versions of this product
Product Code. 17807295
  • 90.20 EUR / Each
Estimated Shipment: 25-02-2025
Description
  • Objective Lens compatible with SMZ160.
  • Not compatible with standard stands.
  • Available in sizes 0.35x with WD = 287 mm, 0.5x with WD = 165 mm, 0.75x with WD = 120 mm, 1.5x with WD = 47 mm and 2.0 with WD = 30 mm.
Specifications
Specifications
SMZ160 Objective
Increase zoom range of SMZ-160
Achromatic
5 years
47 mm
SMZ-160
1.5X
47 mm