missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Objectives, Stereo for SMZ-161/171
Suitable stereo objectives for SMZ-161 and SMZ-171 Microscopes. Motic™ Objectives, Stereo for SMZ-161/171 are precision-crafted for superior quality and durability.
112.00 EUR
Product Code | Brand | Description | Magnification Power | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Magnification Power | Price | Quantity & Availability | ||||
15415452
|
Motic™
1101001703671 |
WD: 56.3mm | 1.5X |
112.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15425442
|
Motic™
1101001703651 |
WD: 301mm | 0.3X |
112.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15465442
|
Motic™
1101001703661 |
WD: 142.7mm | 0.63X |
112.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15495442
|
Motic™
1101001703811 |
WD: 128.6mm | 0.75X |
112.00 EUR
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
Description
- Suitable for SMZ-161 and SMZ-171 Microscopes.
Specifications
Stereo | |
SMZ-161/171 Microscopes |