Learn More
asecos™ Oil Sorbents Cushions
The Oil Sorbents Cusions are water repellant for the safe absorption of oils, petrol, diesel, etc.
Brand:  asecos™ 9932
Additional Details : Weight : 6.00000kg
 View more versions of this product
Product Code. 17896546
  • 89.04 EUR / Pack of 10
Estimated Shipment: 23-07-2024
Description
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibres, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifications
Specifications
White
Oil Sorbent Pillow
460.00 mm
460.00 mm
6 kg
Heavy-duty Cloth
18.11 in.
18.11 in.
13.23 lb.