Learn More
Omadacycline tosylate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Omadacycline tosylate acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline tosylate possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline tosylate can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
Specifications
Specifications
| Chemical Name or Material | Omadacycline tosylate |
| CAS | 1075240-43-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C36H48N4O10S |
| Quantity | 5 mg |
| Synonym | PTK 0796 tosylateAmadacycline tosylate |
| Solubility Information | H2O : 12.5 mg/mL (17.15 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.OS(=O)(C5=CC=C(C)C=C5)=O |
| Molecular Weight (g/mol) | 728.85 |
| Show More |