Learn More
Omadacycline tosylate, MedChemExpress
Product Code. 16472347
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16472347
Brand: Medchem Express HY14865B
MedChemExpress Omadacycline (PTK 0796) tosylate, a first-in-class orally active aminomethylcycline antibacterial, is a member of the tetracycline class of antibiotics.
Omadacycline tosylate acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline tosylate possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline tosylate can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
Specifications
Chemical Name or Material Omadacycline tosylate
CAS 1075240-43-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C36H48N4O10S
Quantity 50 mg
Synonym PTK 0796 tosylateAmadacycline tosylate
Solubility Information H2O : 12.5 mg/mL (17.15 mM; Need ultrasonic)
SMILES O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.OS(=O)(C5=CC=C(C)C=C5)=O
Molecular Weight (g/mol) 728.85
Formula Weight 728.85
Percent Purity 99.37%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation