missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Omega Nitrex™ Extra Length Nitrile Gloves

Product Code. 12811276
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides full arm protection. Nitritex™ Omega Nitrex™ Extra Length Nitrile Gloves is a shoulder-length, ambidextrous glove with a beaded cuff, smooth surface, good puncture resistance and outstanding chemical resistance.

  • Shoulder length (530mm/21 in.) to provide protection when full-arm coverage is required
  • Nitrile is resistant to a wide range of chemicals
  • Beaded cuff provides easier donning and stability on the arm
  • Smooth surface with excellent puncture resistance
  • Powder-free and latex-free to eliminate Type I latex allergies
  • Easy-access 40-glove dispenser box packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Complies with Glove Standard EN 420:2003 & EN 374 (Parts 1, 2 & 3:2003)
Classification Class I Medical & Cat 3 PPE
Color White
Disposable Single-use
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Controlled Environment Gloves
Size Medium
Sterility Non-sterile
Glove Cuff Style Beaded
Packaging Type Dispenser box
Quantity 40/Bx
Resistant To A broad range of chemicals
Series Omega Nitrex
Length (English) 21 in.
Length (Metric) 53 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'