Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
POA
Specifications
Type | Liquid Chromatography Column |
---|---|
Temperature (Metric) Range | 4 to 40°C |
Pore Size | 10 and 30 μm |
Material | Borosilicate Glass, PTFE, Chemraz™ |
Max. Temperature | 40°C |
Product Code | Brand | Column Capacity | Description | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity | Description | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||
15805424
|
Diba
006EZS-06-05-FF |
0.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 50mm; 1/ea | 6 mm | 50 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835424
|
Diba
006EZS-06-10-FF |
2.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 100mm; 1/ea | 6 mm | 100 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865424
|
Diba
006EZS-06-15-FF |
4.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 150mm; 1/ea | 6 mm | 150 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15895424
|
Diba
006EZS-06-25-FF |
7.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 250mm; 1/ea | 6 mm | 250 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825434
|
Diba
006EZS-06-33-FF |
10.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 330mm; 1/ea | 6 mm | 330 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855434
|
Diba
006EZS-06-40-FF |
12.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 400mm; 1/ea | 6 mm | 440 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885434
|
Diba
006EZS-10-10-FF |
5.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 10 x 100mm; 1/ea | 10 mm | 100 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815444
|
Diba
006EZS-10-15-FF |
9.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 10 x 150mm; 1/ea | 10 mm | 150 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15845444
|
Diba
006EZS-10-25-FF |
17.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 10 x 250mm; 1/ea | 10 mm | 250 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15875444
|
Diba
006EZS-10-33-FF |
23.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 10 x 330mm; 1/ea | 10 mm | 330 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805454
|
Diba
006EZS-10-40-FF |
29.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 10 x 400mm; 1/ea | 10 mm | 400 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835454
|
Diba
006EZS-10-50-FF |
36.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 10 x 500mm; 1/ea | 10 mm | 500 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865454
|
Diba
006EZS-15-10-FF |
12.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 15 x 100mm; 1/ea | 15 mm | 100 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15895454
|
Diba
006EZS-15-15-FF |
21.2 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 15 x 150mm; 1/ea | 15 mm | 150 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825464
|
Diba
006EZS-15-25-FF |
38.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 15 x 250mm; 1/ea | 15 mm | 250 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855464
|
Diba
006EZS-15-33-FF |
53 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 15 x 330mm; 1/ea | 15 mm | 330 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885464
|
Diba
006EZS-15-40-FF |
65.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 15 x 400mm; 1/ea | 15 mm | 400 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815474
|
Diba
006EZS-15-50-FF |
83.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 15 x 500mm; 1/ea | 15 mm | 500 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15845474
|
Diba
006EZS-25-10-FF |
34.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 25 x 100mm; 1/ea | 25 mm | 100 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15875474
|
Diba
006EZS-25-15-FF |
58.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 25 x 150mm; 1/ea | 25 mm | 150 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805484
|
Diba
006EZS-25-25-FF |
108 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 25 x 250mm; 1/ea | 25 mm | 250 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835484
|
Diba
006EZS-25-33-FF |
147.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 25 x 330mm; 1/ea | 25 mm | 330 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865484
|
Diba
006EZS-25-40-FF |
181.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 25 x 400mm; 1/ea | 25 mm | 400 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15895484
|
Diba
006EZS-25-50-FF |
230.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 25 x 500mm; 1/ea | 25 mm | 500 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825494
|
Diba
006EZS-35-15-FF |
115.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 35 x 150mm; 1/ea | 35 mm | 150 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855494
|
Diba
006EZS-35-25-FF |
211.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 35 x 250mm; 1/ea | 35 mm | 250 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885494
|
Diba
006EZS-35-33-FF |
288.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 35 x 330mm; 1/ea | 35 mm | 330 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815504
|
Diba
006EZS-35-40-FF |
356 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 35 x 400mm; 1/ea | 35 mm | 400 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15845504
|
Diba
006EZS-50-25-FF |
440.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 50 x 250mm; 1/ea | 50 mm | 250 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15875504
|
Diba
006EZS-50-33-FF |
600.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 50 x 330mm; 1/ea | 50 mm | 330 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805514
|
Diba
006EZS-50-40-FF |
741.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 50 x 400mm; 1/ea | 50 mm | 400 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835514
|
Diba
006EZS-50-50-FF |
941.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Fixed Endpieces, 50 x 500mm; 1/ea | 50 mm | 500 mm |
POA
Each |
N/A | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- PEEK™ column threads offer outstanding durability
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Liquid Chromatography Column | |
10 and 30 μm | |
40°C |
4 to 40°C | |
Borosilicate Glass, PTFE, Chemraz™ | |
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |