Learn More
Diba Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
POA
Specifications
Type | Liquid Chromatography Column |
---|---|
For Use With (Equipment) | Omnifit™ EZ SolventPlus™ Columns |
Material | PTFE |
Product Code | Brand | Description | Endcapped | Diameter (Metric) Inner | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Endcapped | Diameter (Metric) Inner | Price | Quantity & Availability | |||
15805314
|
Diba
006ESAE-06 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 6.6mm; 1/ea | Adjustable | 6.6 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815314
|
Diba
006ESAE-10 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 10mm; 1/ea | Adjustable | 10 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15825314
|
Diba
006ESAE-15 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 15mm; 1/ea | Adjustable | 15 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15835314
|
Diba
006ESAE-25 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 25mm; 1/ea | Adjustable | 25 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15845314
|
Diba
006ESAE-35 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 35mm; 1/ea | Adjustable | 35 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15855314
|
Diba
006ESAE-50 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 50mm; 1/ea | Adjustable | 50 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15865314
|
Diba
006ESFE-06 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 6.6mm; 1/ea | Fixed | 6.6 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15875314
|
Diba
006ESFE-10 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 10mm; 1/ea | Fixed | 10 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15885314
|
Diba
006ESFE-15 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 15mm; 1/ea | Fixed | 15 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15895314
|
Diba
006ESFE-25 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 25mm; 1/ea | Fixed | 25 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15805324
|
Diba
006ESFE-35 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 35mm; 1/ea | Fixed | 35 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15815324
|
Diba
006ESFE-50 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 50mm; 1/ea | Fixed | 50 mm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Specifications
Liquid Chromatography Column | |
Omnifit™ EZ SolventPlus™ Columns | |
PTFE |