Learn More
Köttermann Open Mobile Cabinet
Product Code. 17302109
Click to view available options
No. of Drawers:
2
3
None
No. of Shelves:
1
None
Dimensions (D x W x H):
500 x 450 x 590 mm
500 x 450 x 740 mm
500 x 600 x 590 mm
500 x 600 x 740 mm
500 x 900 x 590 mm
500 x 900 x 740 mm
516 x 450 x 590 mm
516 x 600 x 590 mm
500 x 1200 x 590 mm
500 x 1200 x 740 mm
Unit Size:
Each
Product Code. 17302109
Brand: Köttermann 31500065
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Open Mobile Cabinet offers versatile storage space with an open-front design.
  • Versatile storage space
  • Mobile with lockable wheels
  • Open-front design provides easy access to stored items
  • Many sizes and configurations available
Specifications
Color White
Cabinet Type Mobile
Dimensions (D x W x H) 500 x 600 x 740 mm
Door Type None
Finish Epoxy Coated
Frame Material Steel
No. of Doors None
No. of Drawers None
No. of Shelves 1
Warranty None
Wheel Type Plastic with Brakes
With Casters Yes
Material Steel