Learn More
Macherey-Nagel™ Optima™ FFAP GC Capillary Column
Product Code. 10716322 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
10 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
Unit Size:
Each
Product Code. 10716322
Brand: Macherey-Nagel™ 726344.30
Only null left
Specifications
Diameter (Metric) 0.5 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Optima™ FFAP
USP Type G1, G2, G38
For Use With (Application) Free Carboxylic Acids Analysis, Free Carboxylic Acids
Temperature Range 240°C to 250°C
Film Thickness 0.5 μm
Length (Metric) 30 m
Quantity 1 Each