Learn More
Corning™ LSE™ Optional Head
Shop All Corning Products
Click to view available options
Holds:
12 x 1.5 to 2 mL Tubes
2 x 50 mL Tubes
4 x 15 mL Tubes
Unit Size:
Each
Description
This is an optional head for tubes, held horizontally, for use with Corning™ LSE™ Vortex Mixer.
Specifications
Specifications
| Holds | 4 x 15 mL Tubes |
| Product Type | Optional Head |
| For Use With (Equipment) | Corning™ LSE™ Vortex Mixer |