Learn More
Thermo Scientific™ Orion™ Single Junction Combination pH Electrodes
Product Code. 10282391
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
U.S. Standard
Type:
Combination pH Electrode
Tip Type:
Flask
Flat Surface
Semi-micro
Standard
Unit Size:
Each
Product Code. 10282391
Brand: Thermo Scientific™ 910500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Conduct routine, small volume or flat surface pH measurement with Thermo Scientific™ Orion™ economy series pH electrodes.
Thermo Scientific™ Orion™ Low Maintenance pH Electrodes provide reliable and accurate measurements of routine samples. 9106BNWP is best suited for field or plant use with general purpose samples, and educational institutes where minimal maintenance is required. 911600 is ideal for samples as small as 0.2mL. 912600 is suited for use with long neck flask. 913600 has a flat sensing bulb good for measurement of flat, soft, moist, semi-solid or small volume samples.
  • Permanently gelled Ag/AgCl electrolyte
  • 9106BNWP has 0 to 14 pH range; all others have , 0 to 12 pH range
  • 0° to 80°C (32° to 176°F) temperature range
  • General purpose and flat surface models: 4.31″ long (11cm) × 0.43″ diameter (12mm) body, 0.62″ diameter (16mm) cap
  • Semimicro Model: 5.93″ long (15cm) × 0.25″ diameter (6mm) body, 0.18″ diameter (10mm) cap
91-06 Model:
  • Waterproof BNC connector
General Purpose, Flask, Semimicro and Flat Surface Models:
  • BNC connector
TRUSTED_SUSTAINABILITY
Specifications
Description Permanently gelled Ag/AgCl electrolyte
Connector Types U.S. Standard
Junction Wick
Material Epoxy
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 80°C
Type Combination pH Electrode
Reference Electrolyte Ag/AgCl
Temperature (English) Operating 32°F to 176°F
Tip Type Standard
Length (English) 4.31 in.
Length (Metric) 11 cm
Length (Metric) Cable 1 m
Diameter (English) Body 0.43 in.
Diameter (English) Cap 0.62 in.
Diameter (Metric) Body 12 mm
Diameter (Metric) Cap 16 mm
Refillable Non-refillable