Learn More
Thermo Scientific™ Orion™ Potassium Electrodes
Measure potassium ions in aqueous solutions simply, accurately and economically.
260.30 EUR - 1045.20 EUR
Specifications
Drift | drift-free potentials |
---|---|
Junction | Free-flowing Sure-Flow junction assures stable |
For Use With (Equipment) | ISE Meters |
Type | Combination Ion Selective Electrode |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10282871
|
Thermo Scientific™
931901 |
Replacement Module for Potassium Half-Cell Electrode 9319BN |
260.30 EUR
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
10410643
|
Thermo Scientific™
9319BN |
Potassium Half Cell Electrode with BNC connector |
888.42 EUR
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
10390832
|
Thermo Scientific™
971901 |
Replacement Module for Potassium Sure-Flow Combination Electrode 9719BNWP |
312.56 EUR
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
10757352
|
Thermo Scientific™
9719BNWP |
Potassium Sure-Flow Combination Electrode with Waterproof BNC connector |
1045.20 EUR
Each |
In Stock
Log in to see stock. |
|||||
Description
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
Features
9719BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 971901.
Potassium Half-Cell Electrode 9319BN (comes with half-cell module 931901) is to be used with the Orion 900200 sure flow double junction reference electrode.
Solutions required
Standards
921906 0.1 M potassium standard, 475 mL bottle
ISA
931911 ISA for potassium measurements,475 mL bottle
Electrode fill solution
900002 Inner chamber filling solution for the double junction Reference electrode, 5 x 60 mL bottles
900065 Optimum Results E electrode filling solution,5 x 60 mL bottles
Items included
9319BN 931901 93 series sensing module for 9319BN. Quantity 2.
931911 ISA for potassium measurements. 60ml 1 bottle.
9719BNWP 971901 97 series sensing module for 9719BNWP. Quantity 2.
900065 Optimum Results E electrode filling solution. 60ml 1 bottle.
Electrode : 12 months from date of purchase
Module: 6 months from date of purchase
Specifications
drift-free potentials | |
Free-flowing Sure-Flow junction assures stable | |
ISE Meters | |
Combination Ion Selective Electrode |