missing translation for 'onlineSavingsMsg'
Learn More

ortho-Phosphoric Acid, 85%, ACS Reagent Grade, Scharlab

Product Code. p-7083581
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 11324144

Brand: Scharlab AC11001000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, in fertilizer compositions, as solvents (for pharmaceutical use), and as an antioxidant (in food industry).

  • Synonyms: Orthophosphoric acid
  • Molecular Formula: H3PO4
  • Molar Mass: 98.00g/mol
  • CAS: 7664-38-2
  • EIN ECS No.: 231-633-2
  • Passes test for substances precipitated with ammonia

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Appearance of Solution Clear and colorless
Melting Point ∼21°C
Boiling Point ∼158°C
Density 1.71g/cm3
Identification Passes Test
Vapor Pressure 2.2 hPa (at 25°C)
Quantity 1 L
Physical Form Liquid
Solubility Information Miscible in water (20°C)
Formula Weight 98.00g/mol
Percent Purity ≥85% (acidimetric)
Grade ACS Reagent
Show More Show Less
compliance-icons
Product Identifier
  • ortho-Phosphoric acid, 85%, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Phosphoric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.