Learn More
Osteopontin Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22952-1-AP-20UL
 View more versions of this product
Product Code. 16842145
  • 145.00 EUR / 20µL
Explore more special offers
Description
Description
Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in maligt glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.
Specifications
Specifications
Osteopontin
Polyclonal
Unconjugated
SPP1
BNSP, Bone sialoprotein 1, BSPI, ETA 1, Nephropontin, OPN, Osteopontin, secreted phosphoprotein 1, SPP 1, SPP1, Urinary stone protein, Uropontin
Rabbit
Antigen Affinity Chromatography
RUO
25353, 6696
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08721, P10451
SPP1
Osteopontin Fusion Protein Ag19216
20 μL
Primary
Human, Rat
Antibody
IgG