missing translation for 'onlineSavingsMsg'
Learn More

OTS514, MedChemExpress

Product Code. 16463606
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463606

Brand: Medchem Express HY18621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OTS514 is a highly potent TOPK inhibitor with an IC50 of 2.6 nM. OTS514 strongly suppresses the growth of TOPK-positive cancer cells. OTS514 induces cell cycle arrest and apoptosis.

OTS514 is a highly potent TOPK inhibitor with an IC50 of 2.6 nM. OTS514 strongly suppresses the growth of TOPK-positive cancer cells. OTS514 induces cell cycle arrest and apoptosis.

Specifications

Chemical Name or Material OTS514
CAS 1338540-63-8
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C21H20N2O2S
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (274.38 mM; Need ultrasonic)
SMILES O=C1NC2=C(C(C3=CC=C([C@@H](C)CN)C=C3)=C(O)C=C2C)C4=C1SC=C4
Molecular Weight (g/mol) 364.46
Formula Weight 364.46
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.