Learn More
Labcon Overfit Caps for Culture Tubes
Click to view available options
Color:
Assorted (Red, Yellow, Blue, Green, Orange)
Blue
Green
Natural
Orange
Purple
Red
Yellow
Closure Size:
10 mm
12 mm
13 mm
16 mm
Unit Size:
Pack of 1000
Pack of 10000
Description
- Designed to easily recap blood drawing tubes, glass test tubes, and plastic culture tubes, Overfit Caps are ideal for protocols that require centrifugation, agitation,
and refrigeration. - Offered in a variety of bright colors, Labcon's Overfit Caps are the ideal alternative to costly parafilm or plug caps that are prone to evaporation.
- Cap includes integral thumb lever for easy opening and vertical ridges for secure grip
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polyethylene USP Class VI resin
- Made in USA
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP |
| Closure Size | 16 mm |
| Pre-sterilized | No |
| Product Type | Overfit Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene USP Class VI |
| Color | Purple |
| For Use With (Application) | Protocols that require centrifugation, agitation, and refrigeration |
| Packaging | 1000/Bag, 10 Bags/Cs. |
| Clean Claims | ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) |
| Show More |