missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, ExpertQ, Scharlab

Product Code. 11348624
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES O.O.OC(=O)C(O)=O
IUPAC Name oxalic acid dihydrate
Molecular Weight (g/mol) 126.06
Grade ExpertQ
Quantity 500 g
compliance-icons compliance-icons
Product Identifier
  • 500GR OXALSYRA-2-HYDRAT PA
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Oxalic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.