Learn More
Oxidation Reagent 0.05 M Iodine in 90/10 Pyridine/Water (v/v), BioSyn™, Honeywell Burdick & Jackson™
Click to view available options
:
Oxidation Reagent 0.05 M Iodine in 90/10 Pyridine/Water (v/v), 4 x 2500 mL
Description
- Specifically processed and purified to exacting specifications, ensure outstanding synthesis efficiency
- Ideal for oligonucleotide synthesis regardless of application
- All solvents and reagents are filtered (0.2 micron for oxidation reagents and 0.1 micron for all other reagents) to minimize particulate contamination
Specifications
Specifications
| Chemical Name or Material | Oxidation reagent |
| Melting Point | −42.2°C |
| CAS | 7553-56-2 |
| Color | Colorless to yellow |
| CAS Max % | 0.013 |
| Boiling Point | 115.25°C (1.013 hPa) |
| Quantity | 4 x 2.5 L |
| Packaging | GL45 Glass Bottle |
| Physical Form | Clear liquid |
| Product Line | BioSyn |
Boiling and melting point temperatures are based off the physical main component.